siRNA Details
| Gene Symbol: Gm5091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046164.1 | 5 | 749 | |||
| GenBank mRNA | AK082876.1 | - | 5 | 750 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5091| Gene Symbol: Gm5091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046164.1 | 5 | 749 | |||
| GenBank mRNA | AK082876.1 | - | 5 | 750 | ||