siRNA Details
| Gene Symbol: 1700055C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040726.1 | 3 | 329 | |||
| GenBank mRNA | AK006792.1 | - | 3 | 330 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV204450.1 | - | 3 | 276 | Chr. 9 - 64038465 - 64041406 [-] on Build GRCm38 | |

1700055C04Rik
Genomic Map