siRNA Details
Gene Symbol: Gm20043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135998.1 | - | 1, 2 | 157 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719935.1 | - | 1, 2 | 153 | Chr. 13 - 15375628 - 15406037 [+] on Build GRCm38 |
