siRNA Details
| Gene Symbol: Arhgap26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017029.1 | - | 4 | 717 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716713.1 | - | 4 | 714 | Chr. 18 - 39291457 - 39307370 [+] on Build GRCm38 | |

Arhgap26
Genomic Map