siRNA Details
| Gene Symbol: Lhx1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038057.1 | 4 | 521 | |||
| GenBank mRNA | AK018772.1 | - | 4 | 498 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lhx1os| Gene Symbol: Lhx1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038057.1 | 4 | 521 | |||
| GenBank mRNA | AK018772.1 | - | 4 | 498 | ||