siRNA Details
| Gene Symbol: 4933407L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037692.1 | 2 | 630 | |||
| GenBank mRNA | AK016730.1 | - | 2 | 629 | ||
| AK037440.1 | - | 1 | 1606 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933407L21Rik| Gene Symbol: 4933407L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037692.1 | 2 | 630 | |||
| GenBank mRNA | AK016730.1 | - | 2 | 629 | ||
| AK037440.1 | - | 1 | 1606 | |||