siRNA Details
| Gene Symbol: 4933407O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016733.1 | - | 1 | 765 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX520778.1 | - | 1 | 70 | Chr. 17 - 80063496 - 80064025 [+] on Build GRCm38 | |

4933407O12Rik
Genomic Map