siRNA Details
| Gene Symbol: 4933408J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045810.1 | 2 | 451 | |||
| GenBank mRNA | AK016739.1 | - | 2 | 454 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716420.1 | - | 2 | 454 | Chr. 10 - 93589419 - 93596255 [+] on Build GRCm38 | |

4933408J17Rik
Genomic Map