siRNA Details
| Gene Symbol: 1700110C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045461.1 | 2 | 237 | |||
| GenBank mRNA | AK007157.1 | - | 2 | 238 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA464730.1 | - | 2 | 203 | Chr. 17 - 10324602 - 10329340 [+] on Build GRCm38 | |

1700110C19Rik
Genomic Map