siRNA Details
| Gene Symbol: 8430423G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040686.1 | 3 | 547 | |||
| GenBank mRNA | AK018436.1 | - | 3 | 549 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8430423G03Rik| Gene Symbol: 8430423G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040686.1 | 3 | 547 | |||
| GenBank mRNA | AK018436.1 | - | 3 | 549 | ||