siRNA Details
| Gene Symbol: 8430423G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040686.1 | 2 | 160 | |||
| GenBank mRNA | AK018436.1 | - | 2 | 161 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720566.1 | - | 2 | 161 | Chr. 5 - 148950250 - 148951658 [+] on Build GRCm38 | |

8430423G03Rik
Genomic Map