siRNA Details
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK157944.1 | - | 1 | 1761 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY592453.1 | - | 1 | 431 | Chr. 2 - 76407891 - 76408353 [+] on Build GRCm38 | |

Osbpl6
Genomic Map