siRNA Details
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004353.1 | - | 2 | 226 | ||
| AK157944.1 | - | 1 | 1005 | |||
| BY704583.1 | - | 2 | 226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl6| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004353.1 | - | 2 | 226 | ||
| AK157944.1 | - | 1 | 1005 | |||
| BY704583.1 | - | 2 | 226 | |||