siRNA Details
| Gene Symbol: 9630009A06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020673.1 | - | 11 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722409.1 | - | 11 | Chr. 15 - 30961455 - 30962878 [-] on Build GRCm38 | ||

9630009A06Rik
Genomic Map