siRNA Details
| Gene Symbol: B230110C06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020961.1 | - | 120 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY727002.1 | - | 1, 2 | 116 | Chr. 14 - 15437626 - 15451834 [+] on Build GRCm38 | |

B230110C06Rik
Genomic Map