siRNA Details
| Gene Symbol: B230110G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020962.1 | - | 4 | 601 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY727005.1 | - | 4 | 601 | Chr. 6 - 138580624 - 138599213 [+] on Build GRCm38 | |

B230110G15Rik
Genomic Map