siRNA Details
| Gene Symbol: Gm31019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079798.1 | - | 1 | 33 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB635491.1 | - | 1 | 33 | Chr. 18 - 40437140 - 40439737 [-] on Build GRCm38 | |

Gm31019
Genomic Map