siRNA Details
| Gene Symbol: Gm12440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110516.1 | 2 | 486 | |||
| GenBank mRNA | AK076933.1 | - | 2 | 487 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12440| Gene Symbol: Gm12440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110516.1 | 2 | 486 | |||
| GenBank mRNA | AK076933.1 | - | 2 | 487 | ||