siRNA Details
| Gene Symbol: Gm12440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110516.1 | 2 | 338 | |||
| GenBank mRNA | AK076933.1 | - | 2 | 339 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615799.1 | - | 2 | 339 | Chr. 3 - 99049822 - 99052802 [+] on Build GRCm38 | |

Gm12440
Genomic Map