siRNA Details
| Gene Symbol: 4933407I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040731.1 | 3 | 586 | |||
| GenBank mRNA | AK076937.1 | - | 3 | 587 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715817.1 | - | 3 | 587 | Chr. 9 - 51914429 - 51926841 [+] on Build GRCm38 | |

4933407I05Rik
Genomic Map