siRNA Details
| Gene Symbol: 4933407I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040731.1 | 3 | 778 | |||
| GenBank mRNA | AK076937.1 | - | 3 | 779 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933407I05Rik| Gene Symbol: 4933407I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040731.1 | 3 | 778 | |||
| GenBank mRNA | AK076937.1 | - | 3 | 779 | ||