siRNA Details
| Gene Symbol: Enpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014707.1 | - | 1 | 124 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA118797.1 | - | 1 | 49 | Chr. 10 - 24638308 - 24638716 [-] on Build GRCm38 | |

Enpp1
Genomic Map