siRNA Details
| Gene Symbol: Gm13546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045895.1 | 2, 3 | 195 | |||
| GenBank mRNA | AK041746.1 | - | 2, 3 | 169 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB638452.1 | - | 2, 3 | 166 | Chr. 2 - 58164003 - 58176810 [+] on Build GRCm38 | |

Gm13546
Genomic Map