siRNA Details
| Gene Symbol: Gm13546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045895.1 | 4 | 933 | |||
| NR_045896.1 | 2 | 600 | ||||
| GenBank mRNA | AK041746.1 | - | 4 | 907 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13546| Gene Symbol: Gm13546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045895.1 | 4 | 933 | |||
| NR_045896.1 | 2 | 600 | ||||
| GenBank mRNA | AK041746.1 | - | 4 | 907 | ||