siRNA Details
| Gene Symbol: Gm13648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC048384.1 | - | 2 | 1352 | ||
| BG868839.1 | - | 1 | 163 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13648| Gene Symbol: Gm13648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC048384.1 | - | 2 | 1352 | ||
| BG868839.1 | - | 1 | 163 | |||