siRNA Details
| Gene Symbol: 2700008G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012221.1 | - | 1 | 134 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711398.1 | - | 1 | 134 | Chr. 14 - 63664448 - 63665437 [+] on Build GRCm38 | |

2700008G24Rik
Genomic Map