siRNA Details
| Gene Symbol: 2700008G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012221.1 | - | 1 | 1591 | ||
| BC038474.1 | - | 1 | 742 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2700008G24Rik| Gene Symbol: 2700008G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012221.1 | - | 1 | 1591 | ||
| BC038474.1 | - | 1 | 742 | |||