siRNA Details
| Gene Symbol: A930007I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132712.1 | - | 395 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ046753.1 | - | 5 | 395 | Chr. 19 - 29469776 - 29523034 [-] on Build GRCm38 | |

A930007I19Rik
Genomic Map