siRNA Details
| Gene Symbol: Pitpnm2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045369.1 | 6 | 1427 | |||
| GenBank mRNA | AK016253.1 | - | 6 | 1426 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pitpnm2os1| Gene Symbol: Pitpnm2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045369.1 | 6 | 1427 | |||
| GenBank mRNA | AK016253.1 | - | 6 | 1426 | ||