siRNA Details
| Gene Symbol: Pitpnm2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045369.1 | 1 | 95 | |||
| GenBank mRNA | AK016253.1 | - | 1 | 95 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715864.1 | - | 1 | 95 | Chr. 5 - 124229725 - 124236252 [+] on Build GRCm38 | |

Pitpnm2os1
Genomic Map