siRNA Details
| Gene Symbol: 4930568G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040480.1 | 5 | 938 | |||
| GenBank mRNA | AK016251.1 | - | 5 | 939 | ||
| AK036349.1 | - | 567 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930568G15Rik| Gene Symbol: 4930568G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040480.1 | 5 | 938 | |||
| GenBank mRNA | AK016251.1 | - | 5 | 939 | ||
| AK036349.1 | - | 567 | ||||