siRNA Details
| Gene Symbol: 4930568G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040480.1 | 5 | 786 | |||
| GenBank mRNA | AK016251.1 | - | 5 | 787 | ||
| AK036349.1 | - | 415 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB629215.1 | - | 2 | 407 | Chr. 1 - 165066637 - 165068108 [-] on Build GRCm38 | |

4930568G15Rik
Genomic Map