siRNA Details
Gene Symbol: 4930568G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040480.1 | 2, 3 | 151 | |||
GenBank mRNA | AK016251.1 | - | 2, 3 | 152 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930568G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040480.1 | 2, 3 | 151 | |||
GenBank mRNA | AK016251.1 | - | 2, 3 | 152 |