siRNA Details
| Gene Symbol: Gm38569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042846.1 | - | 1 | 23 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB640063.1 | - | 1 | 21 | Chr. 7 - 25584225 - 25593243 [+] on Build GRCm38 | |

Gm38569
Genomic Map