siRNA Details
| Gene Symbol: Gm5083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045285.1 | 5 | 787 | |||
| GenBank mRNA | AK042843.1 | - | 5 | 788 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5083| Gene Symbol: Gm5083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045285.1 | 5 | 787 | |||
| GenBank mRNA | AK042843.1 | - | 5 | 788 | ||