siRNA Details
| Gene Symbol: 4930482G09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108044.1 | 1 | 85 | |||
| GenBank mRNA | AK015610.1 | - | 1 | 87 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715160.1 | - | 1 | 87 | Chr. 3 - 153147960 - 153152108 [-] on Build GRCm38 | |

4930482G09Rik
Genomic Map