siRNA Details
| Gene Symbol: 4930482G09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108044.1 | 3 | 793 | |||
| GenBank mRNA | AK015610.1 | - | 3 | 795 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930482G09Rik| Gene Symbol: 4930482G09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108044.1 | 3 | 793 | |||
| GenBank mRNA | AK015610.1 | - | 3 | 795 | ||