siRNA Details
| Gene Symbol: AI314760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049818.1 | - | 1 | 677 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB426398.2 | - | 1 | 125 | Chr. 18 - 61586062 - 61586757 [+] on Build GRCm38 | |

AI314760
Genomic Map