siRNA Details
| Gene Symbol: AI314760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049818.1 | - | 1 | 1066 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI314760| Gene Symbol: AI314760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049818.1 | - | 1 | 1066 | ||