siRNA Details
| Gene Symbol: Sox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031243.1 | - | 1 | 665 | ||
| AK042703.1 | - | 1 | 1085 | |||
| AK086149.1 | - | 1 | 1194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sox5| Gene Symbol: Sox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031243.1 | - | 1 | 665 | ||
| AK042703.1 | - | 1 | 1085 | |||
| AK086149.1 | - | 1 | 1194 | |||