siRNA Details
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031243.1 | - | 1 | 1204 | ||
AK042703.1 | - | 1 | 1624 | |||
AK086149.1 | - | 2 | 1802 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031243.1 | - | 1 | 1204 | ||
AK042703.1 | - | 1 | 1624 | |||
AK086149.1 | - | 2 | 1802 |