siRNA Details
| Gene Symbol: Gm26519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043462.1 | - | 3 | 2022 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm26519| Gene Symbol: Gm26519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043462.1 | - | 3 | 2022 | ||