siRNA Details
| Gene Symbol: D630024D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102312.1 | 4, 5 | 611 | |||
| NR_102314.1 | 3, 4 | 459 | ||||
| GenBank mRNA | AK085420.1 | - | 4, 5 | 589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D630024D03Rik| Gene Symbol: D630024D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102312.1 | 4, 5 | 611 | |||
| NR_102314.1 | 3, 4 | 459 | ||||
| GenBank mRNA | AK085420.1 | - | 4, 5 | 589 | ||