siRNA Details
| Gene Symbol: D630024D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102312.1 | 2 | 295 | |||
| GenBank mRNA | AK085420.1 | - | 2 | 273 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D630024D03Rik| Gene Symbol: D630024D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102312.1 | 2 | 295 | |||
| GenBank mRNA | AK085420.1 | - | 2 | 273 | ||