siRNA Details
| Gene Symbol: D630024D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102310.1 | 1 | 136 | |||
| NR_102311.1 | 1 | 136 | ||||
| NR_102312.1 | 1 | 136 | ||||
| NR_102314.1 | 1 | 136 | ||||
| GenBank mRNA | AK085420.1 | - | 1 | 114 | ||
| AK158188.1 | - | 1 | 137 | |||
| AK158233.1 | - | 1 | 119 | |||

D630024D03Rik
Genomic Map