siRNA Details
| Gene Symbol: 1700127G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007300.1 | - | 1 | 41 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707403.1 | - | 1 | 41 | Chr. 18 - 68474713 - 68475212 [-] on Build GRCm38 | |

1700127G19Rik
Genomic Map