siRNA Details
| Gene Symbol: Gm1715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045333.1 | 4 | 527 | |||
| GenBank mRNA | AK138605.1 | - | 4 | 528 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY724359.1 | - | 4 | 524 | Chr. 9 - 52679866 - 52782829 [+] on Build GRCm38 | |

Gm1715
Genomic Map