siRNA Details
| Gene Symbol: Gm12603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144910.1 | - | 3 | 496 | ||
| BB819745.1 | - | 3 | 320 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY734595.1 | - | 3 | 494 | Chr. 4 - 89050308 - 89084511 [+] on Build GRCm38 | |

Gm12603
Genomic Map