siRNA Details
| Gene Symbol: Gm12603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033533.1 | 1 | 207 | |||
| GenBank mRNA | AK144910.1 | - | 1 | 208 | ||
| BB819745.1 | - | 1 | 32 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12603| Gene Symbol: Gm12603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033533.1 | 1 | 207 | |||
| GenBank mRNA | AK144910.1 | - | 1 | 208 | ||
| BB819745.1 | - | 1 | 32 | |||