siRNA Details
| Gene Symbol: Gm12603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033533.1 | 2 | 237 | |||
| GenBank mRNA | AK144910.1 | - | 2 | 238 | ||
| BB819745.1 | - | 2 | 62 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12603| Gene Symbol: Gm12603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033533.1 | 2 | 237 | |||
| GenBank mRNA | AK144910.1 | - | 2 | 238 | ||
| BB819745.1 | - | 2 | 62 | |||